Skip to Content
Merck
All Photos(1)

Key Documents

SRP4247

Sigma-Aldrich

MIP-1beta human

recombinant, expressed in E. coli, ≥99% (SDS-PAGE), ≥99% (HPLC)

Synonym(s):

CCL4, MIP-1- β, Macrophage inflammatory protein 1-β, Small inducible cytokine A4, T-cell activation protein 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥99% (HPLC)
≥99% (SDS-PAGE)

form

lyophilized

mol wt

~7.6 kDa

packaging

pkg of 10 μg

impurities

endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL4(388372)

General description

MIP-1β (macrophage inflammatory protein-1β), or CCL4L1 (C-C motif chemokine ligand 4 like 1), is a CC chemokine gene localized to human chromosome 17q12. It is a copy number variable gene which shares 95% sequence homology with its non-copy number variable paralogues CCL4.[1][2] MIP-1β is expressed by a wide range of cells including immune cells, such as monocytes, B and T cells, fibroblasts, endothelial, and epithelial cells.[3]

Biochem/physiol Actions

MIP-1β (macrophage inflammatory protein-1β), or CCL4L1 (C-C motif chemokine ligand 4 like 1), interacts with its major G protein-coupled receptors CCR5 and CCR8, and promotes the migration of immune cells, such as monocytes, natural killer (NK) cells, and T cells, maturation of dendritic cells (DCs), T cell and granulocyte activation, and T cell differentiation. It is an age associated inflammatory cytokine, and it shows elevated expression in the monocytes and blood of elderly individuals. MIP-1β plays a wide role in immune responses, and hence, its expression is tightly controlled during T-cell differentiation and activation.[3]

Physical form

Lyophilized without any additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional redundancy of the human CCL4 and CCL4L1 chemokine genes.
Howard OM et al
Biochemical and Biophysical Research Communications, 320(3), 927-931 (2004)
Evolution of haplotypes at CCL3L1/CCL4L1.
Janyakhantikul S et al
Genome Biology, P20 (2010)
MicroRNA-125b modulates inflammatory chemokine CCL4 expression in immune cells and its reduction causes CCL4 increase with age.
Cheng NL
Aging Cell, 14(2), 200-208 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service