Skip to Content
Merck
All Photos(1)

Documents

SRP4178

Sigma-Aldrich

IL-17A from rat

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

Ctla8, IL17, IL17a

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

30 ng/mL

mol wt

~30 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

IL-17A (interleukin 17A) is the first cytokine member of the family composed of IL-17A-F. These cytokines are mainly produced by activated CD4+ (cluster of differentiation) T cells. IL-17A is predominantly synthesized by T-helper (Th)-17 lymphocytes, γδ T cells (γδ-17), cytotoxic CD8+ T cells (Tc17), natural killer T (NKT) cells, and neutrophils.

Biochem/physiol Actions

IL-17A (interleukin 17A) acts as a regulator of immune responses in inflammatory disorders, and acts as a potent pleiotropic proinflammatory cytokine where it promotes the release of other pro-inflammatory cytokines from a variety of cells. This cytokine is linked with pain, and IL-17A and IL-17A+ T cells are found in patients with arthritic pain, and in induced animal models of sciatic nerve, and optic nerve injury with neuropathic pain. This protein is thought to be involved in myocardial remodeling and fibrosis, where it promotes the synthesis of collagens and matrix metalloproteinases (MMPs) and activates cardiomyocyte apoptosis.
Recombinant rat IL-17 produced from Escherichia coli is a homodimeric, non-glycosylated polypeptide containing 268 amino acids and has a molecular weight of 30kDa.

Physical form

Lyophilized (from 10 mM Sodium Citrate, pH 3.0).

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 μg/μL (Note: Gently pipetting to dissolve). The solution can then be diluted into other aqueous buffers.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pengfei Ni et al.
Journal of neuroinflammation, 15(1), 332-332 (2018-12-07)
Perioperative neurocognitive disorders (PND) occur frequently after surgery, especially in aged patients. Surgery-induced neuroinflammation and blood-brain barrier (BBB) dysfunction play a crucial role in the pathogenesis of PND. Interleukin-17A (IL-17A) increases after surgical stress and will be involved in BBB
Interleukin-17A Exacerbates Ferric Chloride-Induced Arterial Thrombosis in Rat Carotid Artery.
Maione F et al
International Journal of Inflammation, 2014, 247503-247503 (2014)
Baraa Noueihed et al.
Frontiers in cell and developmental biology, 9, 630645-630645 (2021-02-09)
Ischemic retinopathies (IRs), such as retinopathy of prematurity and diabetic retinopathy, are characterized by an initial phase of microvascular degeneration that results in retinal ischemia, followed by exaggerated pathologic neovascularization (NV). Mesenchymal stromal cells (MSCs) have potent pro-angiogenic and anti-inflammatory
Wendy Dankers et al.
Arthritis research & therapy, 20(1), 212-212 (2018-09-22)
Despite recent improvements in the treatment of rheumatoid arthritis (RA), an insufficient treatment response and the development of treatment resistance in many patients illustrates the need for new therapeutic strategies. Chronic synovial inflammation could be suppressed by targeting RA synovial
Mitsuaki Ishioka et al.
Digestive diseases and sciences, 62(2), 396-406 (2016-12-04)
Although several types of diet have been used in experimental steatohepatitis models, comparison of gut microbiota and immunological alterations in the gut among diets has not yet been performed. We attempted to clarify the difference in the gut environment between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service