Skip to Content
Merck
All Photos(1)

Key Documents

SRP4038

Sigma-Aldrich

Fgf-2 from mouse

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)

Synonym(s):

FGF-b, Fibroblast Growth Factor-basic, HBGF-2 Prostatropin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

mouse

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

~16.3 kDa

packaging

pkg of 50 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Fgf2(14173)

General description

FGF-2 (fibroblast growth factor 2) is a basic heparin binding growth factor.[1] It is ubiquitously expressed and belongs to the FGF family.[2] It is also referred to as BFGF (basic fibroblast growth factor). The recombinant mouse FGF-2 is a single, non-glycosylated chain containing 145 amino acids and having a molecular weight of 16.3kDa.

Biochem/physiol Actions

FGF-2 (fibroblast growth factor 2, also referred to as BFGF (basic fibroblast growth factor)) stimulates the proliferation of a wide variety of cells including mesenchymal, neuroectodermal and endothelial cells.[3][4][5] It is present in the preimplantation embryos and is important for proper development.[6] It is also responsible for angiogenesis.[2] FGF-2 plays a crucial role in adipogenesis and development of adipose tissues.[7] It is associated with tumor growth and metastasis.[8]

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in sterile water to 100 μg/mL.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biphasic effects of FGF2 on adipogenesis.
Kim S
PLoS ONE, 10, e0120073-e0120073 (2015)
Blocking FGF2 with a new specific monoclonal antibody impairs angiogenesis and experimental metastatic melanoma, suggesting a potential role in adjuvant settings.
de Aguiar RB
Cancer Letters, 371, 151-160 (2016)
Binding of FGF2 to FGFR2 in an autocrine mode in trophectoderm cells is indispensable for mouse blastocyst formation through PKC-p38 pathway.
Yang J
Cell Cycle, 14, 3318-3330 (2015)
Yongyi Wang et al.
PloS one, 7(7), e40691-e40691 (2012-08-01)
In fertile women, the endometrium undergoes regular cycles of tissue build-up and regression. It is likely that uterine stem cells are involved in this remarkable turn over. The main goal of our current investigations was to identify slow-cycling (quiescent) endometrial
Interleukin-1? induces fibroblast growth factor 2 expression and subsequently promotes endothelial progenitor cell angiogenesis in chondrocytes.
Chien SY
Clinical Science, 130, 667-681 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service