Skip to Content
Merck
All Photos(1)

Documents

SRP3332

Sigma-Aldrich

M-CSF rat

Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

CSF-1, MGI-IM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

≤5.0 ng/mL ED50

mol wt

36.2 kDa

packaging

pkg of 10 μg

impurities

tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... CSF1(78965)

General description

Macrophage colony stimulating factor (M-CSF) is a potent hematopoietic factor produced by a variety of cells including lymphocytes, monocytes, fibroblasts, endothelial cells, myoblasts and osteoblasts. The gene encoding this protein is localized on rat chromosome 2q34. The human protein is reactive in murine systems, but the murine molecule exhibits no activity on human cells. Recombinant rat M-CSF is a 36.2kDa homodimeric protein containing two 155 amino acid polypeptide subunits.

Biochem/physiol Actions

Macrophage colony stimulating factor (M-CSF) is a key regulator of cellular proliferation, differentiation, and survival of blood monocytes, tissue macrophages and their progenitor cells. M-CSF has been shown to play important roles in modulating dermal thickness, and male and female fertility. M-CSF is clinically used in the treatment of infection, malignancies and atherosclerosis. It facilitates hematopoietic recovery after bone marrow transplantation.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemokines and the tissue-specific migration of lymphocytes.
Kunkel, E.J. and Butcher E.C.
Immunity, 1, 1-4 (2002)
Colony-stimulating factor-1 antibody reverses chemoresistance in human MCF-7 breast cancer xenografts.
Paulus P
Cancer Research, 66(8), 4349-4356 (2006)
Stanley, E.R.
The Cytokine Handbook, 387-418 (1994)
Yuying Tang et al.
Brain, behavior, and immunity, 68, 248-260 (2017-10-31)
Accumulation of microglia occurs in the dorsal horn in the rodent model of chronic post ischemic pain (CPIP), while the mechanism how microglia affects the development of persistent pain largely remains unknown. Here, using a rodent model of CPIP induced
The osteopetrotic mutation toothless (tl) is a loss-of-function frameshift mutation in the rat Csf1 gene: Evidence of a crucial role for CSF-1 in osteoclastogenesis and endochondral ossification.
Van Wesenbeeck L
Proceedings of the National Academy of Sciences of the USA, 99(22), 14303-14308 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service