Skip to Content
Merck
All Photos(1)

Key Documents

SRP3254

Sigma-Aldrich

sRank Ligand from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

OPGL, TNFSF11, TRANCE (TNF-related activation-induced cytokine), soluble Receptor Activator of NFkB Ligand

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€276.00

€276.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€276.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€276.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

10.0-25.0 ng/mL ED50

mol wt

19.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

RANKL and RANK are members of the TNF superfamily of ligands and receptors that play an important role in the regulation of specific immunity and bone turnover. RANK (receptor) was originally identified as a dendritic-cell-membrane protein, which by interacting with RANKL augments the ability of dendritic cells to stimulate naïve T-cell proliferation in a mixed lymphocyte reaction, to promote the survival of RANK + T cells, and to regulate T-cell-dependent immune response. RANKL, which is expressed in a variety of cells including osteoblasts, fibroblasts, activated T-cells and bone marrow stromal cells, is also capable of interacting with a decoy receptor called OPG. Binding of soluble OPG to sRANKL inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage. Recombinant rat sRANKL is a 19.4 kDa polypeptide comprising the TNF homologous region of RANKL (174 amino acid residues).

Biochem/physiol Actions

RANKL and RANK are members of the TNF superfamily of ligands and receptors that play an important role in the regulation of specific immunity and bone turnover. Recombinant rat sRANKL is a 19.4 kDa polypeptide comprising the TNF homologous region of RANKL (174 amino acid residues).

Sequence

PAMMEGSWLD VARRGKPEAQ PFAHLTINAA DIPSGSHKVS LSSWYHDRGW AKISNMTLSN GKLRVNQDGF YYLYANICFR HHETSGSVPA DYLQLMVYVV KTSIKIPSSH NLMKGGSTKN WSGNSEFHFY SINVGGFFKL RAGEEISVQV SNPSLLDPDQ DATYFGAFKV QDID

Physical form

Lyophilized from 5 mM Sodium Phosphate, pH 7.6 + 75 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Lacey et al.
Cell, 93(2), 165-176 (1998-05-06)
The ligand for osteoprotegerin has been identified, and it is a TNF-related cytokine that replaces the requirement for stromal cells, vitamin D3, and glucocorticoids in the coculture model of in vitro osteoclastogenesis. OPG ligand (OPGL) binds to a unique hematopoeitic
D M Anderson et al.
Nature, 390(6656), 175-179 (1997-11-21)
Dendritic cells are rare haematopoietic cells that reside in a number of organs and tissues. By capturing, processing and presenting antigens to T cells, dendritic cells are essential for immune surveillance and the regulation of specific immunity. Several members of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service