Skip to Content
Merck
All Photos(1)

Key Documents

SRP3225

Sigma-Aldrich

MIP-1 gamma (CCL9/10) from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCF18, CCL9/10, MRP2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€276.00

€276.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€276.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

€276.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

11.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CCL9(20308)

General description

MIP-1γ is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. MIP-1γ signals through the CCR1 receptor. MIP-1γ chemoattracts neutrophils and also inhibits colony formation of bone marrow myeloid immature progenitors. MIP-1g contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant murine MIP-1γ is an 11.6 kDa protein containing 101 amino acid residues.

Biochem/physiol Actions

MIP-1g is a CC chemokine found in murine blood and a wide variety of murine tissues, with no known human homolog. Recombinant murine MIP-1g is an 11.6 kDa protein containing 101 amino acid residues.

Sequence

QITHATETKE VQSSLKAQQG LEIEMFHMGF QDSSDCCLSY NSRIQCSRFI GYFPTSGGCT RPGIIFISKR GFQVCANPSD RRVQRCIERL EQNSQPRTYK Q

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youn, B.S., et al.
Cytokine Reference, 1237-1243 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service