Skip to Content
Merck
All Photos(1)

Key Documents

SRP3223

Sigma-Aldrich

MEC (CCL28) from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCK-1, CCL28, Mucosae-associated Epithelial Chemokine

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€260.00

€260.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€260.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€260.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

12.6 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... CCL28(56838)

General description

MEC or chemokine (C-C motif) ligand 28 (CCL28) is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon.[1] MEC contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant murine MEC is a 12.6kDa protein containing 111 amino acid residues.

Biochem/physiol Actions

Chemokine (C-C motif) ligand 28 (CCL28) or MEC signals through the CCR10 receptor and chemoattracts resting CD4, CD8 T-cells, IgA-secreting plasma cells and eosinophils. It has been shown to exhibit antimicrobial activity against bacteria and fungi.[1][2]

Sequence

SEAILPMASS CCTEVSHHVS GRLLERVSSC SIQRADGDCD LAAVILHVKR RRICISPHNR TLKQWMRASE VKKNGRENVC SGKKQPSRKD RKGHTTRKHR TRGTHRHEAS R

Physical form

Lyophilized from 10 mM NaCitrate, pH 4.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The antimicrobial activity of CCL28 is dependent on C-terminal positively-charged amino acids
Wilson E
European Journal of Immunology (2010)
CCL28 induces mucosal homing of HIV-1-specific IgA-secreting plasma cells in mice immunized with HIV-1 virus-like particles
Rainone V
PLoS ONE (2011)
J Pan et al.
Journal of immunology (Baltimore, Md. : 1950), 165(6), 2943-2949 (2000-09-07)
Mucosae-associated epithelial chemokine (MEC) is a novel chemokine whose mRNA is most abundant in salivary gland, with strong expression in other mucosal sites, including colon, trachea, and mammary gland. MEC is constitutively expressed by epithelial cells; MEC mRNA is detected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service