Skip to Content
Merck
All Photos(1)

Key Documents

SRP3206

Sigma-Aldrich

IL-17F from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€329.00

€329.00


Usually ships in 2 weeks.


Select a Size

Change View
25 μG
€329.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€329.00


Usually ships in 2 weeks.

biological source

mouse

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

30 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... IL17F(257630)

General description

IL-17F, a member of the IL-17 family of structurally related cytokines, has been shown to stimulate proliferation and activation of T-cells and PBMCs. IL-17F also regulates cartilage matrix turnover and inhibits angiogenesis. Recombinant murine IL-17F is a disulfide-linked homodimer of 30 kDa, consisting of two 134 amino acid residue chains.

Biochem/physiol Actions

IL-17F, a member of the IL-17 family of structurally related cytokines, has been shown to stimulate proliferation and activation of T-cells and PBMCs. Recombinant murine IL-17F is a disulfide-linked homodimer of 30 kDa, consisting of two 134 amino acid residue chains.

Sequence

MRKNPKAGVP ALQKAGNCPP LEDNTVRVDI RIFNQNQGIS VPREFQNRSS SPWDYNITRD PHRFPSEIAE AQCRHSGCIN AQGQEDSTMN SVAIQQEILV LRREPQGCSN SFRLEKMLLK VGCTCVKPIV HQAA

Physical form

Lyophilized from 10 mM Sodium Acetate, pH 5.0.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T A Moseley et al.
Cytokine & growth factor reviews, 14(2), 155-174 (2003-03-26)
Interleukin-17 (IL-17) is a pro-inflammatory cytokine secreted by activated T-cells. Recently discovered related molecules are forming a family of cytokines, the IL-17 family. The prototype member of the family has been designated IL-17A. Due to recent advances in the human
Zhenhua Luo et al.
Inflammation, 38(3), 978-986 (2014-11-12)
The objective of this study was to investigate the expression level, clinical significance, and possible regulating role of IL-17F in patients of chronic periodontitis. Periodontal local tissues were obtained from chronic periodontitis (CP) and healthy controls (HC) for real-time PCR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service