Skip to Content
Merck
All Photos(1)

Key Documents

SRP3185

Sigma-Aldrich

VEGF-D human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

FIGF, Vascular Endothelial Growth Factor-D

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€329.00

€329.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€329.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€329.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

20.0-22.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FIGF(2277)

General description

VEGFD (vascular endothelial growth factor D), or c-fos-induced growth factor, is a member of the VEGF family of growth factors, and is composed of VEGF-homology domain, receptor binding domains, and pro-peptides in both terminals. It is secreted in the extracellular space in a full length form with both C- and N-terminal pro-peptides, which are cleaved off to produce the mature VEGFD form.[1] It is an angiogenesis-related factor.[2]
Recombinant human VEGF-D is a 26.2 kDa non-disulfide linked homodimeric protein consisting of two 117 amino acid polypeptide chains. Due to glycosylation the protein migrates as a 20.0-22.0 kDa band under non-reducing condition.

Application

VEGFD (vascular endothelial growth factor D) human has been used in for the treatment of buffalo luteal cell culture to determine the effect of VEGFD on the expression of lymphatic endothelial marker (LYVE1), thereby investigating the role of VEGFD in luteal lymphangeogenesis during different stages of estrous cycle.[3]

Biochem/physiol Actions

Mature VEGFD (vascular endothelial growth factor D) interacts with VEGFR-3, a tyrosine kinase receptor, with high affinity. This receptor predominantly resides on adult lymphatic endothelium and participates in lymphangiogenesis. VEGFD also interacts with Nrp-2 (neuropilin), a non-tyrosine kinase receptor, which functions as a lymphangiogenesis-associated factor. This factor promotes tumor lymphangiogenesis and hence, lymph node metastasis in various cancers, such as ovarian cancer.[1] In patients with CRC (colorectal cancer), higher expression of VEGFD is linked with resistance to bevacizumab, and has potential as a biomarker for predicting the benefit of bevacizumab in CRC chemotherapy.[2]

Sequence

FAATFYDIET LKVIDEEWQR TQCSPRETCV EVASELGKST NTFFKPPCVN VFRCGGCCNE ESLICMNTST SYISKQLFEI SVPLTSVPEL VPVKVANHTG CKCLPTAPRH PYSIIRR

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela C García et al.
Zygote (Cambridge, England), 26(3), 242-249 (2018-06-09)
SummaryThe mammalian oviduct plays a pivotal role in the success of early reproductive events. The urokinase plasminogen activator system (uPAS) is present in the bovine oviduct and is involved in extracellular matrix remodelling through plasmin generation. This system can be
VEGF-D-induced draining lymphatic enlargement and tumor lymphangiogenesis promote lymph node metastasis in a xenograft model of ovarian carcinoma.
Du LC et al
Reproductive Biology and Endocrinology, 12, 14-14 (2014)
Vascular endothelial growth factor D expression is a potential biomarker of bevacizumab benefit in colorectal cancer.
Weickhardt AJ et al
British Journal of Cancer, 113(1), 37-45 (2015)
Expression and localization of locally produced growth factors regulating lymphangiogenesis during different stages of the estrous cycle in corpus luteum of buffalo (Bubalus bubalis).
Ali I et al
Theriogenology, 81(3), 428-436 (2014)
Himadri Roy et al.
FEBS letters, 580(12), 2879-2887 (2006-04-25)
Angiogenesis is the process by which new blood vessels are formed from existing vessels. The vascular endothelial growth factors (VEGFs) are considered as key molecules in the process of angiogenesis. The VEGF family currently includes VEGF-A, -B, -C, -D, -E

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service