Skip to Content
Merck
All Photos(1)

Key Documents

SRP3146

Sigma-Aldrich

Prokineticin-2 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

PK2, PROK2, Protein Bv8 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€325.00

€325.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€325.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€325.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.8 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PROK2(60675)

General description

Prokineticin-2 (PK2) is a cysteine-rich secreted protein that is expressed in the testis and in lower levels of the small intestine. PK2 regulates various biological functions including gastrointestinal motility, angiogenesis and circadiam rhythms. It is closely related to EG-VEGF (Prokineticin-1) and binds to two orphan B-protein-coupled receptors termed PK-R1 and PK-R2. Recombinant human Prokineticin-2 is an 8.8 kDa protein consisting of 81 amino acid residues and ten cysteine residues that potentially form five intra-molecular disulfide bonds.It also contains completely conserved N-terminal hexapeptide sequence that is involved in biological activity.[1] PK2 is encoded by the gene mapped to human chromosome 3p13.[2]

Biochem/physiol Actions

Prokineticin-2 (PK2) plays a vital role in regulation of various biological functions, such as gastrointestinal motility, angiogenesis and circadian rhythms. It is closely related to EG-VEGF (endocrine gland-derived vascular endothelial growth factor)/prokineticin-1 (PK1). PK1 and PK2 exert their function by interacting with two G protein-coupled receptors PKR1 and PKR2, respectively.[1][2][3] PK2 is a candidate gene for the pathogenesis of mood disorders.[2]

Sequence

AVITGACDKD SQCGGGMCCA VSIWVKSIRI CTPMGKLGDS CHPLTRKVPF FGRRMHHTCP CLPGLACLRT SFNRFICLAQ K

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prokineticin 2 and circadian clock output.
Zhou Q Y and Cheng M Y
FEBS Journal, 272(22), 5703-5709 (2005)
Possible association of prokineticin 2 receptor gene (PROKR2) with mood disorders in the Japanese population.
Kishi T, et al.
Neuromolecular Medicine, 11(2), 114-122 (2009)
The prokineticins: a novel pair of regulatory peptides.
Zhou Q Y.
Molecular Interventions, 6(6), 330-330 (2006)
Clayton M Bullock et al.
Molecular pharmacology, 65(3), 582-588 (2004-02-24)
Prokineticins are cysteine-rich secreted proteins that regulate diverse biological processes, including gastrointestinal motility, angiogenesis, and circadian rhythms. Two closely related G protein-coupled receptors that mediate signal transduction of prokineticins have recently been cloned. The structural elements required for prokineticins' bioactivities
Takatoshi Soga et al.
Biochimica et biophysica acta, 1579(2-3), 173-179 (2002-11-13)
Recent studies have identified two novel biofunctional proteins, termed prokineticin 1/EG-VEGF and prokineticin 2, which were mammalian homologues of mamba MIT1 and frog Bv8. Prokineticins have been demonstrated to exert their physiological functions through G-protein coupled receptors (GPCRs). In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service