Skip to Content
Merck
All Photos(1)

Key Documents

SRP3129

Sigma-Aldrich

NT-4 human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

Neurotrophic 4/5 (NT-4/NT-5), Neurotrophin-4

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€329.00

€329.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€329.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€329.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

potency

20-50 ng/mL ED50

mol wt

14.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NTF4(4909)

General description

NT-4 is a neurotrophic factor structurally related to b-NGF, BDNF, and NT-3. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-4 is expressed in the prostate, thymus, placenta and skeletal muscle. NT-4 can signal through the LNGFR and trkB receptors and promotes the survival of peripheral sensory sympathetic neurons. Recombinant human NT-4 is a noncovalently linked homodimer, of two 14.0 kDa polypeptide monomers (260 total amino acid residues).

Biochem/physiol Actions

NT-4 is a neurotrophic factor structurally related to b-NGF, BDNF, and NT-3. Recombinant human NT-4 is a noncovalently linked homodimer, of two 14.0 kDa polypeptide monomers (260 total amino acid residues).

Sequence

GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Klein et al.
Neuron, 8(5), 947-956 (1992-05-01)
Neurotrophin-4 is a novel member of the nerve growth factor family of neurotrophins recently isolated from Xenopus and viper DNA. We now report that the Xenopus NT-4 protein (XNT-4) can mediate some of its biological properties through gp145trkB, a murine
N Y Ip et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(7), 3060-3064 (1992-04-01)
Nerve growth factor, brain-derived neurotrophic factor, and neurotrophin-3 (NT-3) are the three members of the neurotrophin family known to exist in mammals. Recently, a fourth neurotrophin (designated neurotrophin-4 or NT-4), which shares all of the features found in the mammalian

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service