Skip to Content
Merck
All Photos(1)

Documents

SRP3120

Sigma-Aldrich

NANOG TAT human

recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

36.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NANOG(79923)

General description

Nanog is a regulatory protein that is associated with undifferentiated pluripotent cells. The expression of Nanog, which is suppressed in all adult tissues, is restricted to embryonic stem cells and to certain pluripotent cancer cells. Decreased expression of Nanog is strongly correlated with cell differentiation. Nanog, most likely, acts as an intracellular regulator, which helps maintain pluripotency and self renewal via a STAT3 independent pathway. The introduction of Nanog, along with Sox2, Oct4, Lin28, into primary human fibroblasts was sufficient to confer a pluripotent state upon the fibroblast genome. The reprogrammed cells thus obtained resemble ESC in morphology and gene expression. Protein transduction using TAT fusion proteins represents an alternative methodology for introducing transcription factors into primary as well as transformed cells. Recombinant human Nanog-TAT is a 36.2 kDa protein, which is synthesized as a 304 amino acid polypeptide plus a 13- residue C-terminal TAT peptide.

Biochem/physiol Actions

Nanog is a regulatory protein that is associated with undifferentiated pluripotent cells. Recombinant human Nanog-TAT is a 36.2 kDa protein, which is synthesized as a 304 amino acid polypeptide plus a 13- residue C-terminal TAT peptide.

Sequence

MSVDPACPQS LPCFEASDCK ESSPMPVICG PEENYPSLQM SSAEMPHTET VSPLPSSMDL LIQDSPDSST SPKGKQPTSA ENSVAKKEDK VPVKKQKTRT VFSSTQLCVL NDRFQRQKYL SLQQMQELSN ILNLSYKQVK TWFQNQRMKS KRWQKNNWPK NSNGVTQKAS APTYPSLYSS YHQGCLVNPT GNLPMWSNQT WNNSTWSNQT QNIQSWSNHS WNTQTWCTQS WNNQAWNSPF YNCGEESLQS CMQFQPNSPA SDLEAALEAA GEGLNVIQQT TRYFSTPQTM DLFLNYSMNM QPEDVGGYGR KKRRQRRR

Physical form

Lyophilized from 5 mM Sodium Citrate pH 3.0 + 100mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam H Hart et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 230(1), 187-198 (2004-04-27)
The murine Nanog gene, a member of the homeobox family of DNA binding transcription factors, has been shown recently to maintain pluripotency of embryonic stem cells. We have used a sequence homology and expression screen to identify and clone the
Kaoru Mitsui et al.
Cell, 113(5), 631-642 (2003-06-06)
Embryonic stem (ES) cells derived from the inner cell mass (ICM) of blastocysts grow infinitely while maintaining pluripotency. Leukemia inhibitory factor (LIF) can maintain self-renewal of mouse ES cells through activation of Stat3. However, LIF/Stat3 is dispensable for maintenance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service