Skip to Content
Merck
All Photos(1)

Key Documents

SRP3112

Sigma-Aldrich

MEC (CCL28) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCK-1, CCL28, Mucosae-associated Epithelial Chemokine

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€336.00

€336.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
€336.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€336.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

12.3 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL28(56477)

General description

MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. MEC signals through the CCR10 receptor and chemoattracts resting CD4, CD8 T-cells and eosinophils. MEC contains six cysteines including the four highly conserved cysteine residues present in CC chemokines. Recombinant human MEC is a 12.3 kDa protein containing 108 amino acid residues.

Biochem/physiol Actions

MEC is a secreted CC chemokine expressed primarily by epithelial cells of the bronchioles, salivary gland, mammary gland and colon. Recombinant human MEC is a 12.3 kDa protein containing 108 amino acid residues.

Sequence

SEAILPIASS CCTEVSHHIS RRLLERVNMC RIQRADGDCD LAAVILHVKR RRICVSPHNH TVKQWMKVQA AKKNGKGNVC HRKKHHGKRN SNRAHQGKHE TYGHKTPY

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Pan et al.
Journal of immunology (Baltimore, Md. : 1950), 165(6), 2943-2949 (2000-09-07)
Mucosae-associated epithelial chemokine (MEC) is a novel chemokine whose mRNA is most abundant in salivary gland, with strong expression in other mucosal sites, including colon, trachea, and mammary gland. MEC is constitutively expressed by epithelial cells; MEC mRNA is detected
Eric J Kunkel et al.
Immunity, 16(1), 1-4 (2002-02-05)
Tissue-selective trafficking of memory and effector T and B lymphocytes is mediated by unique combinations of adhesion molecules and chemokines. The discovery of several related epithelial-expressed chemokines (TECK/CCL25 in small intestine, CTACK/CCL27 in skin, and MEC/CCL28 in diverse mucosal sites)
Emese Zsiros et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(12), 2840-2850 (2015-02-26)
Chemokines are implicated in T-cell trafficking. We mapped the chemokine landscape in advanced stage ovarian cancer and characterized the expression of cognate receptors in autologous dendritic cell (DC)-vaccine primed T cells in the context of cell-based immunotherapy. The expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service