Skip to Content
Merck
All Photos(1)

Key Documents

SRP3105

Sigma-Aldrich

LEC/NCC-4 (CCL16) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CCL16, LCC-1, LMC, Liver-Expressed Chemokine, MTN-1, NCC-4

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€341.00

€341.00


Usually ships in 2 weeks.


Select a Size

Change View
20 μG
€341.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€341.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

11.2 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL16(6360)

General description

LEC is a CC chemokine that can signal through the CCR8 and CCR1 receptors. It is expressed in the liver, spleen, and thymus. LEC is chemotactic towards monocytes and lymphocytes but not neutrophils. Recombinant human LEC is an 11.2 kDa protein containing 97 amino acid residues, including the four conserved cysteine residues present in CC chemokines.

Biochem/physiol Actions

LEC is a CC chemokine that can signal through the CCR8 and CCR1 receptors. It is expressed in the liver, spleen, and thymus. Recombinant human LEC is an 11.2 kDa protein containing 97 amino acid residues, including the four conserved cysteine residues present in CC chemokines.

Sequence

QPKVPEWVNT PSTCCLKYYE KVLPRRLVVG YRKALNCHLP AIIFVTKRNR EVCTNPNDDW VQEYIKDPNL PLLPTRNLST VKIITAKNGQ PQLLNSQ

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C L Tsou et al.
The Journal of experimental medicine, 188(3), 603-608 (1998-08-04)
Hemofiltrate C-C chemokine (HCC)-1 is a recently cloned C-C chemokine that is structurally similar to macrophage inflammatory protein (MIP)-1alpha. Unlike most chemokines, it is constitutively secreted by tissues and is present at high concentrations in normal human plasma. Also atypical
J A Hedrick et al.
Blood, 91(11), 4242-4247 (1998-05-30)
We have identified and characterized a human beta (CC) chemokine, designated HCC-4, that is most closely related to HCC-1 and which demonstrates chemotactic activity for monocytes. Northern analysis of multiple tissue blots and of activated monocytes mRNA shows expression of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service