Skip to Content
Merck
All Photos(1)

Key Documents

SRP3103

Sigma-Aldrich

LAG-1 (CCL4L1) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

ACT-2, CCL4L1, HC21, Lymphocyte Activation Gene-1, T-cell activiation-2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€244.00

€244.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€244.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€244.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

7.7 kDa

packaging

pkg of 20 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CCL4L1(9560)

General description

LAG-1 is CC chemokine that signals through the CCR5 receptor. LAG-1 is identical to MIP-1beta (ACT II isotype) except for two amino acid substitutions; arginine for histidine at position 22 and serine for glycine at position 47 of the mature protein. LAG-1 chemoattracts monocytes, and exhibits activity as an HIV suppressive factor. Recombinant human LAG-1 is a 7.7 kDa protein containing 69 amino acid residues.

Biochem/physiol Actions

LAG-1 is CC chemokine that signals through the CCR5 receptor. Recombinant human LAG-1 is a 7.7 kDa protein containing 69 amino acid residues.

Sequence

APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRSKQV CADPSESWVQ EYVYDLELN

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Baixeras et al.
Molecular immunology, 27(11), 1091-1102 (1990-11-01)
Using subtractive hybridization of a cDNA library we have identified a human gene, termed LAG-1 (for "Lymphocyte Activation Gene-1"). This cDNA codes for a 69 amino-acid polypeptide which belongs to a new class of recently described proteins secreted by activated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service