Skip to Content
Merck
All Photos(1)

Key Documents

SRP3078

Sigma-Aldrich

IL-16 (121aa) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

LCF (Lymphocyte Chemoattractant Factor)

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€329.00

€329.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€329.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€329.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

12.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL16(3603)

General description

IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells and CD4+ monocytes and eosinophils. Analysis by gel filtration suggests that, under physiological conditions, hIL-16 exists predominantly as a noncovalently linked multimer, but that some IL-16 may exist as a monomer. However, only the multimeric form appears to possess chemotactic activity, suggesting that receptor cross-linking may be required for activity. IL-16 also induces expression of IL-2 receptor (IL-2R) and MHC class II molecules on CD4 + T cells. Human and murine IL-16 show significant cross-species reactivity. Recombinant human IL-16 is a 12.4 kDa protein consisting of 121 amino acid residues.

Biochem/physiol Actions

IL-16 is a CD8+ T cell-derived cytokine that induces chemotaxis of CD4+ T cells and CD4+ monocytes and eosinophils. Recombinant human IL-16 is a 12.4 kDa protein consisting of 121 amino acid residues.

Sequence

SAASASAASD VSVESTAEAT VCTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETVQPG DEILQLGGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QSKETTAAGD S

Physical form

Lyophilized from 5 mM Sodium Phosphate, pH 5.5 + 0.5 mM DTT.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Waldman, T.A., et al.
Cytokine Reference, 214-223 (2001)
W W Cruikshank et al.
Journal of immunology (Baltimore, Md. : 1950), 157(12), 5240-5248 (1996-12-15)
We have recently described the cDNA and predicted protein structure of a natural soluble CD4 ligand, IL-16. IL-16 is chemotactic for CD4+ T cells and induces functional IL-2 receptors in CD4+ T cells. The binding of IL-16 to CD4 results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service