Skip to Content
Merck
All Photos(1)

Key Documents

SRP3042

Sigma-Aldrich

FGF-acidic human

Animal-component free, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

ECGF-beta, FGF-1, Fibroblast Growth Factor-acidic, HBGF-1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€407.00

€407.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€407.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€407.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

≥2x10^6 units/mg

mol wt

15.8 kDa

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FGF1(2246)

General description

FGF-acidic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-acidic is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. FGF-acidic has the ability to signal through all the FGF receptors. Recombinant human FGF-acidic is a 15.8 kDa protein consisting of 140 amino acid residues.

Biochem/physiol Actions

FGF-acidic is one of 23 known members of the FGF family. Recombinant human FGF-acidic is a 15.8 kDa protein consisting of 140 amino acid residues.

Sequence

MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D

Physical form

Lyophilized from 5 mM Sodium Phosphate, pH 8.0 + 100 mM NaCl.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 5 mM Sodium Phosphate, pH 8.0 to a concentration of <0.5 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a stabilizer (example 5% Trehalose) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fibroblast growth factors.
Ornitz, D.M., et al.
Genome Biology, 2, 30051-300512 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service