Skip to Content
Merck
All Photos(1)

Key Documents

SRP3026

Sigma-Aldrich

sDLL-4 human

recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Synonym(s):

Delta-like protein 4, Drosophila Delta homolog 4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€336.00

€336.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
25 μG
€336.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€336.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in HEK 293 cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

potency

>1.5 μg/mL (
)

mol wt

54.0 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... DLL4(54567)

General description

Human sDLL4 comprises the extracellular signaling domain of DLL, a member of a structurally-related family of single-pass type I trans-membrane proteins that serve as ligands for Notch receptors. DLL4 functions to specifically activate the Notch-1 and Notch-4 receptors. The Notch signaling pathway regulates endothelial-cell differentiation, proliferation and apoptosis, and is essential for the development, maintenance and remodeling of the vascular system. Targeted deletion of the DLL4 gene in mice resulted in severe vascular defects and death before birth.[1] Up-regulation of DLL4 expression has been implicated in the vascular development of certain tumors. Recombinant human sDLL4 is a 54.0 kDa glycoprotein containing 498 amino-acid residues. DLL4 is a membrane-bound notch ligand that belongs to the d protein family. Apart from the membrane-bound region, it consists of extracellular growth factor (EGF)-like domains and a receptor-binding DSL domain. DLL4 is selectively expressed in the endothelial cells and may act as a regulator of blood vessel biology.[2][3]

Biochem/physiol Actions

Human sDLL4 comprises the extracellular signaling domain of DLL, a member of a structurally-related family of single-pass type I trans-membrane proteins that serve as ligands for Notch receptors. Recombinant human sDLL4 is a 54.0 kDa glycoprotein containing 498 amino-acid residues.

Sequence

SGVFQLQLQE FINERGVLAS GRPCEPGCRT FFRVCLKHFQ AVVSPGPCTF GTVSTPVLGT NSFAVRDDSS GGGRNPLQLP FNFTWPGTFS LIIEAWHAPG DDLRPEALPP DALISKIAIQ GSLAVGQNWL LDEQTSTLTR LRYSYRVICS DNYYGDNCSR LCKKRNDHFG HYVCQPDGNL SCLPGWTGEY CQQPICLSGC HEQNGYCSKP AECLCRPGWQ GRLCNECIPH NGCRHGTCST PWQCTCDEGW GGLFCDQDLN YCTHHSPCKN GATCSNSGQR SYTCTCRPGY TGVDCELELS ECDSNPCRNG GSCKDQEDGY HCLCPPGYYG LHCEHSTLSC ADSPCFNGGS CRERNQGANY ACECPPNFTG SNCEKKVDRC TSNPCANGGQ CLNRGPSRMC RCRPGFTGTY CELHVSDCAR NPCAHGGTCH DLENGLMCTC PAGFSGRRCE VRTSIDACAS SPCFNRATCY TDLSTDTFVC NCPYGFVGSR CEFPVGLP

Physical form

Lyophilized from 1x PBS, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular cloning of delta-4, a new mouse and human Notch ligand
T Yoneya
Biochemistry, 129 (2001)
Haploinsufficiency of delta-like 4 ligand results in embryonic lethality due to major defects in arterial and vascular development
Nicholas W
Proceedings of the National Academy of Sciences of the USA (2004)
G Thurston et al.
British journal of cancer, 99(8), 1204-1209 (2008-10-02)
Tumour angiogenesis has become an important target for antitumour therapy, with most current therapies aimed at blocking the VEGF pathway. However, not all tumours are responsive to VEGF blockers, and some tumours that are responsive initially may become resistant during
Dll4, a novel Notch ligand expressed in arterial endothelium
J R Shutter
Genes & Development, 14 (2000)
Minhong Yan et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 13(24), 7243-7246 (2007-12-21)
Intense research efforts have been focused toward the identification of regulators of angiogenesis and the development of antiangiogenesis-based cancer therapies. The approval of anti-vascular endothelial growth factor (VEGF) monoclonal antibody (bevacizumab) for use in colorectal and lung cancer provides clinical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service