Skip to Content
Merck
All Photos(1)

Key Documents

SRP2163

Sigma-Aldrich

Estrogen Receptor human

recombinant, expressed in insect cells, ≥80% (SDS-PAGE)

Synonym(s):

DKFZp686N23123, ER, ESR, ESRA, Era, NR3A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

biological source

human

recombinant

expressed in insect cells

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~68 kDa

packaging

pkg of 4 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

200 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ESR1(2099)

General description

The gene ESR1 (estrogen receptor-alpha) encodes a member of the superfamily of nuclear hormone receptors that are essential for the functioning of female reproductive system. The gene is mapped to human chromosome 6q25.

Biochem/physiol Actions

Several members of the nuclear receptor family are directly associated with human malignancies including breast cancer, prostate cancer and leukemia. The pathogenesis of each of these diseases is underpinned by the activities of a member of the superfamily; estrogen receptor-alpha (ER alpha) in breast cancer, androgen receptor (AR) in prostate cancer, and retinoic acid receptor alpha (RAR alpha) in acute promyelocytic leukemia. Estrogen receptors (ER) are members of the superfamily of nuclear hormone receptors (2, 3) whose activity is required for the normal function of the female reproductive system. Two isoforms of estrogen receptor (ER α and ER β) have been described. They function as ligand-dependent transcriptional activators. The biological functions downstream of ER result from altered expression of direct transcriptional targets as well as secondary effects mediated by biological activities of direct targets. In the mammary gland, estrogen receptors regulate normal epithelial cell development and differentiation through their well-documented effects on transcription. Estrogens have long been known to have mitogenic functions in breast cancer cell lines and in breast tumors. Selective estrogen receptor modulatory compounds (SERMs), which bind directly to ER, can block the growth stimulatory function of estrogens.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Estrogen receptor alpha (ESR1) gene amplification is frequent in breast cancer.
Holst F
Nature Genetics, 39, 655-660 (2007)
Hyo Jin Gim et al.
European journal of medicinal chemistry, 85, 107-118 (2014-08-01)
A series of azaisoflavone analogs were designed and synthesized and their transactivation activities and binding affinities for ERα and ERβ were investigated. Among these compounds, 2b and 3a were the most potent with 6.5 and 1.1 μM of EC50, respectively. Molecular
Molecular mechanisms of action of steroid/thyroid receptor superfamily members.
M J Tsai et al.
Annual review of biochemistry, 63, 451-486 (1994-01-01)
Activating ESR1 mutations in hormone-resistant metastatic breast cancer.
Robinson DR
Nature Genetics, 45, 1446-1451 (2013)
Juan-Juan Yin et al.
International journal of nanomedicine, 10, 4717-4730 (2015-08-08)
Breast cancer is a leading killer of women worldwide. Cyclodextrin-based estrogen receptor-targeting drug-delivery systems represent a promising direction in cancer therapy but have rarely been investigated. To seek new targeting therapies for membrane estrogen receptor-positive breast cancer, an estrogen-anchored cyclodextrin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service