Skip to Content
Merck
All Photos(1)

Key Documents

SRP2138

Sigma-Aldrich

LXR β isoform, ligand binding domain (211-461), GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

LXR-b, LXRB, NER, NER-I, RIP15, UNR

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€363.00

€363.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€363.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

€363.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~55.8 kDa

packaging

pkg of 10 μg

concentration

300 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NR1H2(7376)

Related Categories

Biochem/physiol Actions

Liver X receptors (LXRs) are nuclear receptors that regulate the metabolism of cholesterol and bile acids. There are two subtypes of LXRs, LXRa and LXRb. LXRb is preferentially expressed in liver, small intestine, kidney and spleen. In contrast, LXRa expression is ubiquitous. The genomic structure and the promoter regions of the two LXR genes contain specific regulatory sites, which suggest that LXRs may have physiological roles in the immune system. Like other nuclear receptors, LXRs heterodimerize with retinoid X receptor (RXR) for function. LXRs are activated by naturally occurring oxysterols and regulate the expression of target genes, including ATP binding cassette transporter 1 (ABC1), ATP binding cassette transporter 8 (ABC8) and cholesterol ester transfer protein (CETP). LXRb expressed in livers of LXRa knockout mice does not compensate for the loss of LXRa. In addition, LXRb, but not LXRa, is also able to activate transcription of a reporter gene, which contains a specific direct repeat separated by 1 bp (DR1) element in the promoter, suggesting that LXRb may have different biological functions.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D J Peet et al.
Current opinion in genetics & development, 8(5), 571-575 (1998-10-31)
The liver X receptors (LXRs) are a family of transcription factors that were first identified as orphan members of the nuclear receptor superfamily. The identification of a specific class of oxidized derivatives of cholesterol as ligands for the LXRs has
R Apfel et al.
Molecular and cellular biology, 14(10), 7025-7035 (1994-10-01)
The steroid/hormone nuclear receptor superfamily comprises several subfamilies of receptors that interact with overlapping DNA sequences and/or related ligands. The thyroid/retinoid hormone receptor subfamily has recently attracted much interest because of the complex network of its receptor interactions. The retinoid
P J Willy et al.
Genes & development, 9(9), 1033-1045 (1995-05-01)
We have identified a new retinoid response pathway through which 9-cis retinoic acid (9cRA) activates transcription in the presence of LXR alpha, a member of the nuclear receptor superfamily. LXR alpha shows a specific pattern of expression in visceral organs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service