Skip to Content
Merck
All Photos(1)

Key Documents

SRP2125

Sigma-Aldrich

RXRα, ligand binding domain, (200-462), GST tagged human

recombinant, expressed in E. coli, ≥80% (SDS-PAGE)

Synonym(s):

FLJ00280, FLJ00318, FLJ16020, FLJ16733, MGC102720, NR2B1

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€272.00

€272.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€272.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

€272.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥80% (SDS-PAGE)

form

frozen liquid

mol wt

~56.5 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RXRA(6256)

Biochem/physiol Actions

Retinoid X receptor (RXR) serves as a promiscuous heterodimerization partner for many nuclear receptors through the identity box, a 40-amino acid subregion within the ligand binding domain (LBD). RXR partners include thyroid hormone receptors (TRs), retinoic acid receptors (RARs), peroxisome proliferator-activated receptor, several constitutive active orphan nuclear receptors (e.g. nuclear growth factor I-B), oxysterol receptors, and constitutive androstane receptors. RXRs also form homodimers to mediate the effects of 9-cis-retinoic acid (9-cRA). Depending on these protein-protein interactions, RXR-containing complexes have distinct ligand-dependent and constitutive functions. The LBD is functionally complex and mediates ligand binding, receptor homo- and heterodimerization, repression of transcription in the absence of ligand, and ligand-dependent activation of transcription. Hormone binding to the structurally conserved LBD of the RXR triggers a conformational change that principally affects the conserved C-terminal transactivation helix H12 involved in transcriptional activation. Coactivators directly recruited by liganded receptors include members of the steroid receptor coactivator/p160 family such as SRC-1, transcriptional intermediary factor 2/glucocorticoid receptor interacting protein 1, and RAC3/activator of thyroid and retinoic acid receptors/amplified in breast cancer 1.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Nakashima et al.
Science (New York, N.Y.), 284(5413), 479-482 (1999-04-16)
The cytokines LIF (leukemia inhibitory factor) and BMP2 (bone morphogenetic protein-2) signal through different receptors and transcription factors, namely STATs (signal transducers and activators of transcription) and Smads. LIF and BMP2 were found to act in synergy on primary fetal
The RXR heterodimers and orphan receptors.
D J Mangelsdorf et al.
Cell, 83(6), 841-850 (1995-12-15)
The nuclear receptor superfamily: the second decade.
D J Mangelsdorf et al.
Cell, 83(6), 835-839 (1995-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service