Skip to Content
Merck
All Photos(1)

Key Documents

SRP2112

Sigma-Aldrich

TFIIB, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

TF2B, TFIIB

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€402.00

€402.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€402.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€402.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~61.2 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

400 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... GTF2B(2959)

Biochem/physiol Actions

The transcription factor IIB (TFIIB) is an essential factor for transcription by RNA polymerase II. TFIIB has been shown to be required for selective binding of RNA polymerase II, TFIIF and TFIID-DNA complex, and for specifying the start site of transcription. Human TFIIB is a single, 35 kDa polypeptide, homologous to yeast factor, the product of the SUA7 gene.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

General initiation factors for RNA polymerase II.
R C Conaway et al.
Annual review of biochemistry, 62, 161-190 (1993-01-01)
S Malik et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(21), 9553-9557 (1991-11-01)
Transcription factor TFIIB is a ubiquitous factor required for transcription initiation by RNA polymerase II. Previous studies have suggested that TFIIB serves as a bridge between the "TATA"-binding factor (TFIID) and RNA polymerase II during preinitiation complex assembly and, more
The general transcription factors of RNA polymerase II.
G Orphanides et al.
Genes & development, 10(21), 2657-2683 (1996-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service