Skip to Content
Merck
All Photos(1)

Key Documents

SRP2106

Sigma-Aldrich

TATA box binding protein,GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

GTF2D, GTF2D1, HDL4, MGC117320, MGC126054, MGC126055, SCA17, TFIID

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€356.00

€356.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€356.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€356.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~64.4 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

350 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TBP(6908)

Biochem/physiol Actions

TBP is the core protein of the transcription factor IID complex, the multiprotein DNA-binding factor that coordinates activities necessary for initiation of transcription by RNA polymerase II.
The TATA-binding protein (TBP) is believed to function as an essential factor of the general transcription machinery and to be involved in transcription by all three eukaryotic RNA polymerases (pol I, II, and III). TBP specifically binds to the TATA element at the promoter region and interacts with numerous transcription factors, including TBP-associated factors (TAFs), activators, and some tumor suppressor proteins.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Purification of his-tagged proteins in non-denaturing conditions suggests a convenient method for protein interaction studies.
A Hoffmann et al.
Nucleic acids research, 19(22), 6337-6338 (1991-11-25)
M Horikoshi et al.
Cell, 54(7), 1033-1042 (1988-09-23)
The mammalian activator protein ATF stimulates transcription from the adenovirus E4 promoter by binding to multiple upstream promoter and enhancer elements. DNAase footprint analyses have revealed that there are cooperative interactions between ATF and TFIID (the mammalian TATA factor) when
TBP, a universal eukaryotic transcription factor?
N Hernandez
Genes & development, 7(7B), 1291-1308 (1993-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service