Skip to Content
Merck
All Photos(1)

Key Documents

SRP2072

Sigma-Aldrich

HDAC3 human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

HD3, RPD3, RPD3-2

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€475.00

€475.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
5 μG
€475.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.26

€475.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~50 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

300 μg/mL

color

colorless to clear

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HDAC3(8841)

Biochem/physiol Actions

The nucleosomal histones can be modified through reversible acetylation by histone acetyltransferases (HATs) and deacetylases (HDACs). Acetylation and deacetylation of histone proteins alters chromosome structure and affects transcription factor access to DNA. HDAC3 has an open reading frame of 428 amino acids. HDAC3 shares 53 % amino acid identity with HDAC1 and 52 % with HDAC2. Observations indicate that class II HDACs regulate transcription by bridging the enzymatically active NCOR2-HDAC3 complex and select transcription factors. All HDACs were found to be ubiquitously expressed in immune and non-immune tissues. In human myeloid leukemia THP-1 cells, HDAC3 transfection resulted in increased size, aberrant nuclear morphology and cell cycle G2/M cell accumulation. Functional activity of the expressed HDAC3 protein was confirmed in alpha-HDAC3 antibody immunoprecipitates by a histone deacetylase assay. Study suggests the participation of HDACs in cell cycle progression and activation.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Yang et al.
The Journal of biological chemistry, 272(44), 28001-28007 (1997-11-05)
Several human cDNAs encoding a histone deacetylase protein, HDAC3, have been isolated. Analysis of the predicted amino acid sequence of HDAC3 revealed an open reading frame of 428 amino acids with a predicted molecular mass of 49 kDa. The HDAC3
F Dangond et al.
Biochemical and biophysical research communications, 242(3), 648-652 (1998-02-17)
The nucleosomal histones can be modified through reversible acetylation by histone acetyltransferases (HATs) and deacetylases (HDACs). HATs induce nucleosomal relaxation and allow DNA-binding by transcriptional activators. HDACs from corepressor complexes which negatively regulate cell growth. However, the HDAC inhibitors butyrate
Rachel Fellows et al.
Nature communications, 9(1), 105-105 (2018-01-11)
The recently discovered histone post-translational modification crotonylation connects cellular metabolism to gene regulation. Its regulation and tissue-specific functions are poorly understood. We characterize histone crotonylation in intestinal epithelia and find that histone H3 crotonylation at lysine 18 is a surprisingly
Hemangi Patil et al.
The Journal of biological chemistry, 291(7), 3158-3172 (2015-12-15)
Histone deacetylase 3 (HDAC3) and linker histone H1 are involved in both chromatin compaction and the regulation of mitotic progression. However, the mechanisms by which HDAC3 and H1 regulate mitosis and the factors controlling HDAC3 and H1 activity during mitosis
David E Wright et al.
Antioxidants & redox signaling, 29(4), 377-388 (2017-11-10)
Thioredoxin reductase 1 (TrxR1) is a cancer target and essential selenoprotein that defends the cell against reactive oxygen species and regulates cellular signaling and redox pathways. Previous cell-based studies correlated TrxR1 acetylation with modulated cellular reduction activity, yet the function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service