Skip to Content
Merck
All Photos(1)

Key Documents

SRP2048

Sigma-Aldrich

RAR, α human

recombinant, expressed in insect cells, ≥70% (SDS-PAGE)

Synonym(s):

NR1B1, RAR

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€536.00

€536.00


Usually ships in 2 weeks.


Select a Size

Change View
5 μG
€536.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€536.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in insect cells

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~52.4 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

1000 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RARA(5914)

Biochem/physiol Actions

Several members of the nuclear receptor family are directly associated with human malignancies including breast cancer, prostate cancer and leukemia. The pathogenesis of each of these diseases is underpinned by the activities of a member of the superfamily; estrogen receptor-α (ERα) in breast cancer, androgen receptor (AR) in prostate cancer, and retinoic acid receptor alpha (RAR alpha) in acute promyelocytic leukemia. Retinoic acid receptors are important in the regulation of growth and differentiation of epithelial tissues, embryonic and central nervous system development and hematopoiesis. Retinoids mediate their effect by two classes of nuclear receptor proteins, the retinoic acid receptors (RARs) and the retinoid X receptors (RXRs), that each consists of three isotypes (α, β, and γ) encoded in separate genes. Upon dimerization with RXR, RARs can bind to specific enhancer sequences in the DNA, so-called retinoic acid response elements (RAREs), resulting in transcriptional activation of target genes in the presence of ligand. The retinoic acid receptor alpha gene is the target of chromosomal rearrangements in all cases of acute promyelocytic leukemia (APL). RARα is a negative regulator of promyelocyte differentiation when not complexed with RA, and stimulates this differentiation when bound to RA.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen M Hart
Biological research, 35(2), 295-303 (2002-11-06)
Nuclear receptors comprise a family of transcription factors that regulate gene expression in a ligand dependent manner. They can activate or repress target genes by binding directly to DNA response elements as homo- or hetero-dimers or by binding to other
M Leid et al.
Trends in biochemical sciences, 17(10), 427-433 (1992-10-01)
Complexity in the retinoid signalling system arises from a combination of several forms of retinoic acid, multiple cytoplasmic binding proteins and nuclear receptors, and the existence of polymorphic retinoic acid response elements. Additional diversity appears to be generated by heterodimeric
Nonsteroid nuclear receptors: what are genetic studies telling us about their role in real life?
P Kastner et al.
Cell, 83(6), 859-869 (1995-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service