Skip to Content
Merck
All Photos(1)

Key Documents

SRP2025

Sigma-Aldrich

p52 human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

DFS70, LEDGF, MGC74712, PAIP, p52

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€522.00

€522.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€522.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€522.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~39 kDa

packaging

pkg of 10 μg

concentration

600 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PSIP1(11168)

Biochem/physiol Actions

The human p52 protein is a non-TAF transcription coactivator that mediates activator-dependent transcription by RNA polymerase II. The function of p52 is through interactions with transcriptional activators and the basal transcription machinery. In addition, p52 may also interact with several cellular proteins including the transcription coactivator PC4, the essential splicing factor ASF/SF2 and the nuclear protein nucleolin.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ge
Nucleic acids research, 28(2), e3-e3 (1999-12-22)
Protein-protein interactions have been widely used to study gene expression pathways and may be considered as a new approach to drug discovery. Here I report the development of a universal protein array (UPA) system that provides a sensitive, quantitative, multi-purpose
H Ge et al.
Molecular cell, 2(6), 751-759 (1999-01-14)
Increasing evidence suggests that pre-mRNA splicing can take place cotranscriptionally in vivo. However, insight into how these two processes are linked has been lacking. Here, we describe that a novel transcriptional coactivator, p52, interacts not only with transcriptional activators and
H Ge et al.
The EMBO journal, 17(22), 6723-6729 (1998-11-21)
Transcriptional activation in human cell-free systems containing RNA polymerase II and general initiation factors requires the action of one or more additional coactivators. Here, we report the isolation of cDNAs encoding two novel human transcriptional coactivators (p52 and p75) that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service