Skip to Content
Merck
All Photos(2)

Key Documents

SRP0733

Sigma-Aldrich

PCTK2 (CDK17)/CYCLINY, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€430.00

€430.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€430.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€430.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

15-25 nmol/min·mg (radiometric)

mol wt

~64 kDa (Cyclin Y)
~88 kDa (PCTK2)

concentration

0.1 μg/μL

accession no.

NM_002595
NM_145012

shipped in

dry ice

storage temp.

−70°C

Gene Information

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant full-length human PCTK2 (CDK17) and Cyclin Y (2-end) were co-expressed by Baculovirus in Sf9 insect cells using N-terminal GST tags. PCTK2 (CDK17) or cyclin-dependent kinase 17 belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. PCTK2 has similarity to a rat protein that is thought to play a role in terminally differentiated neurons (1). The identification of a large family of cdc2-related kinases opens the possibility of combinatorial regulation of the cell cycle together with the emerging large family of cyclins (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service