Skip to Content
Merck
All Photos(2)

Key Documents

SRP0716

Sigma-Aldrich

MYLK4, Active

PRECISIO® Kinase, recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€388.00

€388.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€388.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€388.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

specific activity

4.0-6.0 nmol/min·mg (radiometric)

mol wt

~70 kDa

concentration

0.1 μg/μL

accession no.

BC132833

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MYLK4(340156)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant full-length human MYLK4 was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. MYLK4 or myosin light chain kinase family, member 4 is Serine/Threonine protein kinases in which MYLK4 is one of four myosin light chain kinases in a comprehensive analysis of protein kinases encoded by the human genome.

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service