Skip to Content
Merck
All Photos(1)

Key Documents

SRP0715

Sigma-Aldrich

PEAK1, UNACTIVE

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€368.00

€368.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€368.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€368.00


Usually ships in 2 weeks.

recombinant

expressed in baculovirus infected Sf9 cells

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~100 kDa

concentration

0.1 μg/μL

accession no.

NM_024776

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PEAK1(79834)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant human PEAK1 (1127-end) was expressed by Baculovirus in Sf9 insect cells using an N-terminal GST tag. PEAK1 or Pseudopodium-enriched atypical kinase 1 is a protein tyrosine kinase which belongs to NKF3 kinase family and regulates the cytoskeleton and has been shown to be involved in cancer progression (1). PEAK1 promotes anchorage-independent growth in the MDA-MB-435 human cancer cell line and promotes tumor progression of XPA-1 human pancreatic cancer cells when transplanted into the pancreas of nude mice. PEAK1 is highly expressed in spleen, followed by heart, ovary, brain, lung, kidney, and fetal brain (2).

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service