Skip to Content
Merck
All Photos(1)

Key Documents

SRP0697

Sigma-Aldrich

TAU-316 Protein

recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€388.00

€388.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€388.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€388.00


Usually ships in 2 weeks.

recombinant

expressed in E. coli

Assay

70-95% (SDS-PAGE)

form

frozen liquid

mol wt

~39 kDa

concentration

0.2 μg/μL

accession no.

P10636-3

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAPT(4137)

Application

Useful for kinase assay and Western blot.

Biochem/physiol Actions

Recombinant full-length human tag-free Tau-316 was expressed in E. coli cells. Tau-316 encodes the microtubule-associated protein tau (MAPT) whose transcript undergoes complex, regulated alternative splicing, giving rise to several mRNA species. Tau-316 transcripts are differentially expressed in the nervous system, depending on stage of neuronal maturation and neuron type (1). Tau-316 gene mutations have been associated with several neurodegenerative disorders such as Alzheimer′s disease, Pick′s disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy (2).

Sequence

Full Length

Reconstitution

Formulated in 50mM Tris-HCl, pH 7.5, 150 mM NaCl, 0.25 mM DTT, 0.1 mM PMSF, with 25% glycerol.

Storage and Stability

Store in aliquots at -70°C. Avoid freeze/thaw cycles.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service