Skip to Content
Merck
All Photos(1)

Key Documents

SRP0674

Sigma-Aldrich

IL-17RA, FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

CD217, interleukin-17 receptor a

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
€170.00
25 μG
€426.00

€170.00


Estimated to ship on19 May 2025



Select a Size

Change View
5 μG
€170.00
25 μG
€426.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€170.00


Estimated to ship on19 May 2025


recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

62 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IL17RA(23765)

General description

Human secreted interleukin 17 receptor A (IL-17RA) Fc fusion protein, also known as, IL-17 receptor A and CD217, GenBank Accession No. NM_014339, a.a 33-320, fused at the C-terminus to the Fc portion of human IgG1 with a Cterminal Avitag , expressed in a HEK293 cell expression system. The protein is enzymatically biotinylated using Avitag technology. This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service