Skip to Content
Merck
All Photos(1)

Key Documents

SRP0666

Sigma-Aldrich

IDH2, FLAG® -Tag

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

IDH2, Isocitrate Dehydrogenase 2, oxalosuccinate decarboxylase

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€384.00
100 μG
€677.00

€384.00


Estimated to ship on20 May 2025



Select a Size

Change View
50 μG
€384.00
100 μG
€677.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€384.00


Estimated to ship on20 May 2025


recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

52 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... IDH2(3418)

General description

Human isocitrate dehydrogenase 2, also known as IDH2, (GenBank Accession No. NM_002168), a.a. 1-452(end) with C-terminal FLAG-tag, MW = 52 kDa, expressed in a baculovirus infected Sf9 cell expression system.

Physical form

Aqueous solution

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service