Skip to Content
Merck
All Photos(1)

Key Documents

SRP0663

Sigma-Aldrich

CBL-B, HIS- AviTag (Human)

recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in E. coli

form

frozen liquid

mol wt

48 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CBLB(868)

General description

Human Casitas B-lineage lymphoma proto-oncogene b (CBL-B), also known as E3 ubiquitin-protein ligase CBL-B, RING finger protein 56, and SH3-binding protein CBL-B, GenBank Accession No. NM_170662, a.a. 39 - 426 with N-terminal His-tag and N-terminal Avi-tag, expressed in an E. coli expression system. MW=48 kDa.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service