Skip to Content
Merck
All Photos(1)

Key Documents

SRP0565

Sigma-Aldrich

ICOS (CD278), FC Fusion

recombinant, expressed in HEK 293 cells

Synonym(s):

AILIM, CD278, CVID1, Inducible T-cell costimulator and CD antigen 278, activation-inducible lymphocyte immunomediatory molecule

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€434.00

€434.00


Check Cart for Availability


Select a Size

Change View
100 μG
€434.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€434.00


Check Cart for Availability

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

40.2 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ICOS(29851)

General description

Human ICOS, Fc fusion protein, also known as inducible T-cell costimulator and CD antigen 278, activation-inducible lymphocyte immunomediatory molecule, AILIM, CD278, and CVID1. GenBank Accession No. NM_012092, a.a. 21-140 expressed in a HEK293 cell expression system. MW = 40.2 kDa. This protein runs at a higher MW by SDS-PAGE due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service