Skip to Content
Merck
All Photos(1)

Key Documents

SRP0529

Sigma-Aldrich

Mouse PD-L1 (CD274), FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

B7 homolog 1 (B7-H1), CD274, Programmed Cell Death 1 Ligand 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€591.00

€591.00


Estimated to ship on15 April 2025



Select a Size

Change View
50 μG
€591.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€591.00


Estimated to ship on15 April 2025


recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

54 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... CD274(60533)

General description

Mouse secreted Programmed Cell Death 1 Ligand 1 (PD-L1)-Fc fusion protein, also known as CD274 and B7 homolog 1 (B7- H1), GenBank Accession No. NM_021893, a.a. 18-238, fused at the C-terminus to the Fc portion of mouse IgG2a and with a Cterminal Avitag, expressed in a HEK293 cell expression system. This protein was biotinylated using AviTag technology. MW = 54 kDa. Note: this protein runs at an apparent higher molecular weight due to glycosylation.

Physical form

Aqueous solution

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service