Skip to Content
Merck
All Photos(1)

Key Documents

SRP0526

Sigma-Aldrich

B7-1 (CD80), FC Fusion, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

CTLA-4 Counter-Receptor B7.1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€434.00

€434.00


Estimated to ship on05 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€434.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€434.00


Estimated to ship on05 May 2025


Request a Bulk Order

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

52.5 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CD8O(941)

General description

CD80 or B7-1 is expressed in the antigen-presenting cells (APC).[1] The CD80 gene is mapped to human chromosome 3q13.33.[2]
Human secreted B7-1-Fc fusion protein, also known as CD80, with C-terminal Avitag . GenBank Accession No. NM_005191, a.a 35-242 expressed in a HEK293 cell expression system and enzymatically biotin-labeled using Avitag technology. MW = 52.5 kDa (monomer). This protein runs at a higher M.W. by SDS-PAGE due to glycosylation.

Biochem/physiol Actions

CD80 is an efficient costimulatory molecule essential for immune surveillance during carcinogenesis. It inhibits cancer progression.[3] It regulates the T cell activation to control immune response.[1] Cell stress increases the CD80 expression. Reduced expression of CD80 is observed in colon carcinoma. High levels of CD80 stimulate anti-tumor immune responses. Therefore, enhancing CD80 expression in tumors might serve as a potent approach for cancer immunotherapy.[3] Urinary CD80 is recognized as an important biomarker for idiopathic nephrotic syndrome (INS). CD80 might be involved in proteinuria, actin reorganization, and podocyte migration. Its expression in podocytes is associated with minimal change disease (MCD).[1]

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frank A Schildberg et al.
Immunity, 44(5), 955-972 (2016-05-19)
Immune responses need to be controlled for optimal protective immunity and tolerance. Coinhibitory pathways in the B7-CD28 family provide critical inhibitory signals that regulate immune homeostasis and defense and protect tissue integrity. These coinhibitory signals limit the strength and duration
Gabriel Cara-Fuentes et al.
Pediatric nephrology (Berlin, Germany), 35(10), 1887-1896 (2020-05-14)
Urinary CD80 has emerged as potential biomarker in idiopathic nephrotic syndrome (INS). However, its cellular source remains controversial. The aim of the study was to assess whether CD80 is truly expressed by glomerular cells in INS patients during relapse and
Melania Scarpa et al.
Oncoimmunology, 10(1), 1907912-1907912 (2021-04-20)
CD80 is recognized as one of the most potent costimulatory molecules by which immune cells limit cancer progression; however, the current understanding of the regulation of its expression on human tumor cells is limited. The TP53 tumor suppressor plays a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service