Skip to Content
Merck
All Photos(1)

Key Documents

SRP0513

Sigma-Aldrich

MGLL, His-Tag, GST-Tag

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

HUK5, MAGL, MGLL, Monoacylglycerol Lipase, monoglyceride lipase

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€441.00

€441.00


Estimated to ship on29 May 2025



Select a Size

Change View
50 μG
€441.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

€441.00


Estimated to ship on29 May 2025


recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

62 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MGLL(11343)

General description

Human monoglyceride lipase (MGLL), GenBank Accession No. NM_007283, a.a. 2-313(end), with N-terminal His-tag and N-terminal GST-tag, MW=62 kDa, expressed in Sf9 insect cells via a baculovirus expression system.

Physical form

Aqueous solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service