Skip to Content
Merck
All Photos(2)

Key Documents

SRP0478

Sigma-Aldrich

BAZ2B (2054-2168) human

recombinant, expressed in E. coli, ≥84% (SDS-PAGE)

Synonym(s):

WALp4, bromodomain adjacent to zinc finger domain, 2B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€713.00

€713.00


Usually ships in 2 weeks.


Select a Size

Change View
100 μG
€713.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€713.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥84% (SDS-PAGE)

form

aqueous solution

mol wt

15.1 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BAZ2B(29994)

General description

Human Bromodomain adjacent to zinc finger domain, 2B or BAZ2B (GenBank Accession No. NM_013450) amino-acids 2054 – 2168 (end) with N-terminal HIS-tag, MW = 14.3 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan E Arking et al.
PLoS genetics, 7(6), e1002158-e1002158 (2011-07-09)
Sudden cardiac death (SCD) continues to be one of the leading causes of mortality worldwide, with an annual incidence estimated at 250,000-300,000 in the United States and with the vast majority occurring in the setting of coronary disease. We performed
M H Jones et al.
Genomics, 63(1), 40-45 (2000-02-09)
The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. Bromodomain proteins have been identified as integral components of chromatin remodeling complexes and frequently possess histone acetyltransferase activity. Their encoding genes have been identified at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service