Skip to Content
Merck
All Photos(1)

Key Documents

SRP0470

Sigma-Aldrich

TAF1L (1398-1516) GST tag human

recombinant, expressed in E. coli, ≥82% (SDS-PAGE)

Synonym(s):

TAF1 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 210 kDa-like, TAF2A2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€713.00

€713.00


Usually ships in 2 weeks.


Select a Size

Change View
100 μG
€713.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€713.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥82% (SDS-PAGE)

form

aqueous solution

mol wt

40.6 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TAF1L(138474)

General description

Human TAF1 RNA polymerase II, TAT box binding protein (TBP)-associated factor, also known as TAF1L (GenBank accession No. NM_153809) amino acids 1398 – 1516 with N-terminal GST-tag, MW = 40.6 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Jeremy Wang et al.
Human molecular genetics, 11(19), 2341-2346 (2002-09-10)
TAF(II)250, the largest subunit of the general transcription factor TFIID, is expressed from the human X chromosome, at least in somatic cells. In male meiosis, however, the sex chromosomes are transcriptionally silenced, while the autosomes remain active. How then are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service