Skip to Content
Merck
All Photos(1)

Key Documents

SRP0448

Sigma-Aldrich

BRD1 (561-668) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

BRL, Bromodomain containing 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€527.00

€527.00


Usually ships in 2 weeks.


Select a Size

Change View
100 μG
€527.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€527.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

13.7 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... BRD1(23774)

General description

Human Bromodomain containing 1, also known as BRD1 and BRL (GenBank Accession No. NM_014577) amino-acids 561 – 668 with N-terminal HIS-tag, MW = 13.7 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Severinsen et al.
Molecular psychiatry, 11(12), 1126-1138 (2006-08-23)
Linkage studies suggest that chromosome 22q12-13 may contain one or more shared susceptibility genes for schizophrenia (SZ) and bipolar affective disorder (BPD). In a Faeroese sample, we previously reported association between microsatellite markers located at 22q13.31-qtel and both disorders. The
Yuta Mishima et al.
Blood, 118(9), 2443-2453 (2011-07-15)
The histone acetyltransferases (HATs) of the MYST family include TIP60, HBO1, MOZ/MORF, and MOF and function in multisubunit protein complexes. Bromodomain-containing protein 1 (BRD1), also known as BRPF2, has been considered a subunit of the MOZ/MORF H3 HAT complex based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service