Skip to Content
Merck
All Photos(2)

Key Documents

SRP0427

Sigma-Aldrich

PDE2A FLAG tag active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥88% (SDS-PAGE)

Synonym(s):

cGSPDE, phosphodiesterase 2A, cGMP-stimulated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥88% (SDS-PAGE)

form

aqueous solution

mol wt

106 kDa

packaging

pkg of 5 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE2A(5138)

General description

Human PDE2A (GenBank Accession No. NM_002599) amino-acids 2-941 with N-terminal FLAG-tag, MW=106 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergio E Martinez et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(20), 13260-13265 (2002-09-25)
Cyclic nucleotide phosphodiesterases (PDEs) regulate all pathways that use cGMP or cAMP as a second messenger. Five of the 11 PDE families have regulatory segments containing GAF domains, 3 of which are known to bind cGMP. In PDE2 binding of
Joachim Seybold et al.
Blood, 105(9), 3569-3576 (2005-01-15)
The pleiotropic cytokine tumor necrosis factor-alpha (TNF-alpha) and thrombin lead to increased endothelial permeability in sepsis. Numerous studies demonstrated the significance of intracellular cyclic nucleotides for the maintenance of endothelial barrier function. Actions of cyclic adenosine monophosphate (cAMP) and cyclic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service