Skip to Content
Merck
All Photos(2)

Key Documents

SRP0426

Sigma-Aldrich

PDE1B active rat

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE)

Synonym(s):

PDE1B1, PDES1B, phosphodiesterase 1B, calmodulin-dependent

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€518.00

€518.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€518.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€518.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

rat

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

mol wt

89 kDa

packaging

pkg of 10 μg

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

rat ... PDE1B(5153)

General description

Rat PDE1B (GenBank Accession No. NM_022710) full length with N-terminal HIS-GST-tag, MW= 89kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew T Bender et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(2), 497-502 (2004-12-31)
Granulocyte-macrophage colony-stimulating factor (GM-CSF) is a major regulator of monocyte to macrophage differentiation. In both humans and mice, the main phenotype of decreased GM-CSF function is pulmonary proteinosis due to aberrant function of alveolar macrophages. Recently, this cytokine has been
Andrew T Bender et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(2), 460-465 (2006-01-13)
Monocyte-to-macrophage differentiation with the cytokine granulocyte-macrophage colony-stimulating factor induces expression of the cyclic nucleotide phosphodiesterase PDE1B2. However, what role PDE1B2 plays in macrophage biology has not been elucidated. We have addressed this question by inhibiting PDE1B2 induction by using RNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service