Skip to Content
Merck
All Photos(1)

Key Documents

SRP0420

Sigma-Aldrich

RbBP5 human

recombinant, expressed in E. coli, ≥65% (SDS-PAGE)

Synonym(s):

RBQ3, SWD1, retinoblastoma binding protein 5

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€713.00

€713.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€713.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€713.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥65% (SDS-PAGE)

form

aqueous solution

mol wt

60 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RBBP5(5929)

General description

Human retinoblastoma binding protein 5 (RbBP5), (GenBank Accession No. NM_005057), amino acids 2 – 538 (end) with N-terminal His-tag, MW= 60 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cao, F., et al.
PLoS ONE, 5, e14102-e14102 (2012)
Zain Odho et al.
The Journal of biological chemistry, 285(43), 32967-32976 (2010-08-19)
Histone modification is well established as a fundamental mechanism driving the regulation of transcription, replication, and DNA repair through the control of chromatin structure. Likewise, it is apparent that incorrect targeting of histone modifications contributes to misregulated gene expression and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service