Skip to Content
Merck
All Photos(1)

Key Documents

SRP0418

Sigma-Aldrich

WDR5 human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

BIG-3, SWD3, WD Repeat Domain 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€673.00

€673.00


Usually ships in 2 weeks.


Select a Size

Change View
100 μG
€673.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€673.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

37.5 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WDR5(11091)

General description

Human WDR5 (WD Repeat Domain 5), also known as SWD3 or BIG-3 (GenBank Accession No. NM_017588) amino acids 2-334 (end) with N-terminal His-tag, MW=37.5 kDa, expressed in Sf9 insect cells via a Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

WDR5, a complexed protein.
Raymond C Trievel et al.
Nature structural & molecular biology, 16(7), 678-680 (2009-07-07)
Yan-Yi Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 107(2), 815-820 (2010-01-19)
Viral infection causes activation of the transcription factors NF-kappaB and IRF3, which collaborate to induce type I interferons (IFNs) and cellular antiviral response. The mitochondrial outer membrane protein VISA acts as a critical adapter for assembling a virus-induced complex that
Zain Odho et al.
The Journal of biological chemistry, 285(43), 32967-32976 (2010-08-19)
Histone modification is well established as a fundamental mechanism driving the regulation of transcription, replication, and DNA repair through the control of chromatin structure. Likewise, it is apparent that incorrect targeting of histone modifications contributes to misregulated gene expression and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service