Skip to Content
Merck
All Photos(1)

Key Documents

SRP0386

Sigma-Aldrich

NSD1 human

recombinant, expressed in E. coli, ≥63% (SDS-PAGE)

Synonym(s):

KMT3B, STO, nuclear receptor binding SET domain protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€636.00

€636.00


Usually ships in 2 weeks.


Select a Size

Change View
50 μG
€636.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€636.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in E. coli

Assay

≥63% (SDS-PAGE)

form

aqueous solution

mol wt

54 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NSD1(64324)

General description

Human NSD1 (GenBank Accession No. NM_022455), amino-acids 1849-2094 with N-terminal GST tag, MW = 54 kDa, expressed in an E.coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiko Yamada-Okabe et al.
Cell biochemistry and function, 26(2), 146-150 (2007-04-18)
Nuclear receptor-binding SET-domain-containing protein 1 (NSD1), a culprit gene for Sotos syndrome, contains a su(var)3-9, enhancer-of-zeste, trithorax (SET) domain that is responsible for histone methyltransferase activity and other domains such as plant homeodomain (PHD) and proline-tryptophan-tryptophan-proline (PWWP) involved in protein-protein
Pascale Saugier-Veber et al.
Human mutation, 28(11), 1098-1107 (2007-06-15)
Sotos syndrome is an overgrowth syndrome characterized by distinctive facial features, learning difficulties, and macrocephaly with frequent pre- and postnatal overgrowth with advanced bone age. Here, we report on our experience in the molecular diagnostic of Sotos syndrome on 116

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service