Skip to Content
Merck
All Photos(2)

Key Documents

SRP0383

Sigma-Aldrich

EHMT1 human

recombinant, expressed in E. coli, ≥63% (SDS-PAGE)

Synonym(s):

EHMT1, KMT1D

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€488.00

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€488.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥63% (SDS-PAGE)

form

aqueous solution

mol wt

72.4 kDa

packaging

pkg of 50 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... EHMT1(79813)

General description

Human euchromatic histone-lysine N-methyltransferase (EHMT1), also known as G9a-like protein (GLP) or KMT1D, (GenBank Accession No. NM_024757), amino acids 894-1298 (end) with N-terminal GST-tag, MW= 72.4 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bassant Orabi et al.
Heart failure reviews, 23(3), 363-376 (2018-04-24)
Glucagon-like peptide-1 (GLP-1) agonists and dipeptidyl peptidase-4 (DPP-4) inhibitors produce some beneficial and deleterious effects in diabetic patients not mediated by their glycemic lowering effects, and there is a need for better understanding of the molecular basis of these effects.
Levi Todd et al.
The Journal of comparative neurology, 524(1), 74-89 (2015-06-09)
Retinal progenitors in the circumferential marginal zone (CMZ) and Müller glia-derived progenitors have been well described for the eyes of fish, amphibians, and birds. However, there is no information regarding a CMZ and the nature of retinal glia in species
Jing Huang et al.
The Journal of biological chemistry, 285(13), 9636-9641 (2010-02-02)
The tumor suppressor p53 is regulated by numerous post-translational modifications. Lysine methylation has recently emerged as a key post-translational modification that alters the activity of p53. Here, we describe a novel lysine methylation site in p53 that is carried out
Yukihiro Itoh et al.
Scientific reports, 9(1), 767-767 (2019-01-27)
In the context of drug design, C-H···O hydrogen bonds have received little attention so far, mostly because they are considered weak relative to other noncovalent interactions such as O-H···O hydrogen bonds, π/π interactions, and van der Waals interactions. Herein, we
Shinichiro Kato et al.
Cancer discovery, 10(7), 980-997 (2020-04-10)
Epigenetic regulators, when genomically altered, may become driver oncogenes that mediate otherwise unexplained pro-oncogenic changes lacking a clear genetic stimulus, such as activation of the WNT/β-catenin pathway in melanoma. This study identifies previously unrecognized recurrent activating mutations in the G9a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service