Skip to Content
Merck
All Photos(1)

Key Documents

SRP0359

Sigma-Aldrich

NEK6 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

NIMA (never in mitosis gene a)-related kinase 6, SID6-1512

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€534.00

€534.00


Usually ships in 2 weeks.


Select a Size

Change View
10 μG
€534.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€534.00


Usually ships in 2 weeks.

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

40.1 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NEK6(10783)

General description

Human NEK6 (GenBank Accession No. NM_014397), amino acids 1-313 with N-terminal HIS6-tag, MW=40.1 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will phosphorylate 1 nmol NEK6 substrate (FLAKSFGSPNRAYKK) per minute at pH 7.4 and 30°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Jean Yin et al.
The Journal of biological chemistry, 278(52), 52454-52460 (2003-10-18)
The Aspergillus nidulans protein NIMA (never in mitosis, gene A) is a protein kinase required for the initiation of mitosis, whereas its inactivation is necessary for mitotic exit. Here, we demonstrate that human NIMA-related kinase 6 (Nek6) is required for
Christopher Belham et al.
The Journal of biological chemistry, 278(37), 34897-34909 (2003-07-04)
The Nek family of protein kinases in humans is composed of 11 members that share an amino-terminal catalytic domain related to NIMA, an Aspergillus kinase involved in the control of several aspects of mitosis, and divergent carboxyl-terminal tails of varying
Laura O'Regan et al.
The Journal of cell biology, 209(3), 349-358 (2015-05-06)
Hsp70 proteins represent a family of chaperones that regulate cellular homeostasis and are required for cancer cell survival. However, their function and regulation in mitosis remain unknown. In this paper, we show that the major inducible cytoplasmic Hsp70 isoform, Hsp72

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service