Skip to Content
Merck
All Photos(2)

Key Documents

SRP0332

Sigma-Aldrich

FYN A active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥60% (SDS-PAGE)

Synonym(s):

FYN oncogene related to SRC, FGR, YES, p59-FYN

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€517.00

€517.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
10 μG
€517.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€517.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥60% (SDS-PAGE)

form

aqueous solution

mol wt

61 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FYN(2534)

General description

Human FYN A (GenBank Accession No. NM_002037), full length, amino acids 2-537 (end) with N-terminal Flag-tag, MW= 61 kDa, expressed in Sf9 insect cells.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Fomulated in 45 mM Tris-HCl, pH 8.0, 124 mM NaCl, 2.4 mM KCl, 18 mM glutathione, and 10% glycerol.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kathleen E McGrath et al.
Blood, 111(4), 2409-2417 (2007-11-23)
Enucleation is the hallmark of erythropoiesis in mammals. Previously, we determined that yolk sac-derived primitive erythroblasts mature in the bloodstream and enucleate between embryonic day (E)14.5 and E16.5 of mouse gestation. While definitive erythroblasts enucleate by nuclear extrusion, generating reticulocytes
Hyang-Sook Hoe et al.
The Journal of biological chemistry, 283(10), 6288-6299 (2007-12-20)
Dab1 is an intracellular adaptor protein that interacts with amyloid precursor protein (APP) and apoE receptor 2 (apoEr2), increases their levels on the cell surface, and increases their cleavage by alpha-secretases. To investigate the mechanism underlying these alterations in processing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service