Skip to Content
Merck
All Photos(2)

Key Documents

SRP0321

Sigma-Aldrich

JMJD2D active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

KDM4D, lysine (K)-specific demethylase 4D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€488.00

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
100 μG
€488.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

67 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM4D(55693)

General description

Human JMJD2D, also known as KDM4D (GenBank Accession No. NM_018039), amino-acids 2-354 with N-terminal GST tag, MW=67 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masuko Katoh et al.
International journal of oncology, 24(6), 1623-1628 (2004-05-13)
JMJD1 family genes, including JMJD1 (TSGA), TRIP8, and C5orf7 (5qNCA), encode nuclear proteins with Jumonji C (JmjC), TRI8H1 and TRI8H2 domains. Here, we identified JMJD2 family genes. Human genes corresponding to KIAA0677, KIAA0876, KIAA0780 and FLJ10251 cDNAs were designated JMJD2A
Sook Shin et al.
Biochemical and biophysical research communications, 353(4), 973-977 (2007-01-09)
JMJD2A-D belong to the JmjC domain-containing family of histone demethylases. JMJD2D is the most structurally divergent JMJD2 protein as it lacks the PHD and Tudor domains present in JMJD2A-C. Here, we systematically analyzed the histone demethylase specificity of JMJD2 proteins

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service