Skip to Content
Merck
All Photos(2)

Key Documents

SRP0318

Sigma-Aldrich

JMJD1A human

recombinant, expressed in baculovirus infected Sf9 cells, ≥40% (SDS-PAGE)

Synonym(s):

JHDM2A, KDM3A, lysine (K)-specific demethylase 3A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€488.00

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
20 μG
€488.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

€488.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥40% (SDS-PAGE)

form

aqueous solution

mol wt

148.2 kDa

packaging

pkg of 20 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... KDM3A(55818)

General description

Human JMJD1A (also known as KDM3A or JHDM2A), (GenBank Accession No. NM_018433), amino acids 2-1322 (end) with Nterminal FLAG-tag, MW=148.2 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Physical form

Formulated in 40 mM Tris-HCl, pH 8.0, 124 mM NaCl, 2.7 mM KCl, 90 ng/uL Flag peptide, 10% Glycerol, and 3 mM DTT.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo X. et al.
Neoplasma, 52, 153-157 (2011)
Zi-Ming Du et al.
PloS one, 6(4), e19137-e19137 (2011-05-05)
The role of microRNA-155 (miR-155) has been associated with oncogenesis of several human tumors. However the expression pattern of miR-155 has not been investigated in nasopharyngeal carcinoma (NPC). The present study was to assess miR-155 expression pattern and its possible

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service